Merck
  • Kinetics of phosphate absorption in lactating dairy cows after enteral administration of sodium phosphate or calcium phosphate salts.
Kinetics of phosphate absorption in lactating dairy cows after enteral administration of sodium phosphate or calcium phosphate salts.
The British journal of nutrition (2013-04-03)
Walter Grünberg, Paul Dobbelaar, Gerhard Breves
ABSTRACT
Hypophosphataemia is frequently encountered in dairy cows during early lactation. Although supplementation of P is generally recommended, controversy exists over the suitability of oral P supplementation in animals with decreased or absent rumen motility. Since the effects of transruminal P absorption and the reticular groove reflex on the absorption kinetics of P are not well understood, it is unclear in how far treatment efficacy of oral P supplementation is affected by decreased rumen motility. Phosphate absorption was studied in six phosphate-depleted dairy cows fitted with rumen cannulas and treated with test solutions containing either NaH2PO4 or CaHPO4 with acetaminophen. Each animal was treated orally, intraruminally and intra-abomasally in randomised order. Absorption kinetics of P were studied and compared with the absorption kinetics of acetaminophen, a marker substance only absorbed from the small intestine. Intra-abomasal treatment with NaH2PO4 resulted in the most rapid and highest peaks in plasma inorganic P (Pi) concentration. Oral and intraruminal administration of NaH2PO4 resulted in similar increases in plasma Pi concentration from 4 to 7 h in both groups. Treatment with NaH2PO4 caused more pronounced peaks in plasma Pi concentration compared with CaHPO4. Neither transruminal P absorption nor the reticular groove reflex affected P absorption kinetics as determined by comparing plasma concentration–time curves of P and acetaminophen after administration of 1M-phosphate salt solutions. It is concluded that oral treatment with NaH2PO4 but not CaHPO4 is effective in supplementing P in hypophosphataemic cows with adequate rumen motility. Decreased rumen motility is likely to hamper the efficacy of oral phosphate treatment.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, SAJ first grade, ≥98.0
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ACS reagent, 98.0-102.0%
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)