Merck
  • Hand asepsis: the efficacy of different soaps in the removal of bacteria from sterile, gloved hands.
Hand asepsis: the efficacy of different soaps in the removal of bacteria from sterile, gloved hands.
Journal of the American Dental Association (1939) (1986-08-01)
J P Gobetti, M Cerminaro, C Shipman
ABSTRACT
The study showed that washing a gloved hand removed significant amounts of bacteria. If the proper soap or scrub is used, the gloved hand will be free of bacteria. It is suggested that all dental personnel wear gloves to protect themselves and to prevent cross-contamination of other patients.
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from cold water fish skin, 40-50% in H2O
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A