Merck
  • Relationships between glutamate and monoamine metabolites in cerebrospinal fluid and serum in healthy volunteers.
Relationships between glutamate and monoamine metabolites in cerebrospinal fluid and serum in healthy volunteers.
Biological psychiatry (1988-04-01)
G Alfredsson, F A Wiesel, A Tylec
ABSTRACT
The concentrations of homovanillic acid (HVA), 5-hydroxyindoleacetic acid (5-HIAA), 4-hydroxy-3-methoxyphenylglycol (HMPG), and glutamate were determined in cerebrospinal fluid (CSF) and serum in 10 healthy volunteers. The monoamine metabolites were measured by mass fragmentography and the glutamate by high-performance liquid chromatography. The level of glutamate in CSF was low (0.34 +/- 0.14 nmol/ml) in comparison with previously published values. The concentrations of monoamine metabolites in CSF were in close agreement with earlier findings. There were negative correlations between the concentrations of HVA (r = -0.82, p less than 0.01) and 5-HIAA (r = -0.77, p less than 0.01) and glutamate in CSF, but not in serum. The serum levels of HMPG and glutamate were negatively correlated (r = -0.95, p less than 0.001), but not the CSF levels. The HMPG levels in serum and CSF were positively correlated (r = 0.94, p less than 0.001), but not the HVA and the 5-HIAA levels. The serum and CSF levels of glutamate were positively correlated (r = 0.67, p less than 0.05). The results indicate relationships among the metabolism of dopamine, serotonin, and glutamate in the brain and between the metabolism of noradrenaline and glutamate in peripheral tissue.
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
L-Glutamic acid, JIS special grade, ≥99.0%
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland