Merck
  • 1H NMR characterization of normal human cerebrospinal fluid and the detection of methylmalonic acid in a vitamin B12 deficient patient.
1H NMR characterization of normal human cerebrospinal fluid and the detection of methylmalonic acid in a vitamin B12 deficient patient.
NMR in biomedicine (1991-08-01)
F Commodari, D L Arnold, B C Sanctuary, E A Shoubridge
ABSTRACT
1H NMR was used to analyze human cerebrospinal fluid (CSF) from a group of neurological disease controls and from a vitamin B12 deficient patient. The spectra were acquired at either 7.06 or 9.40 T at ambient temperature with CSF freeze dried and reconstituted in 2H2O. 3-Trimethylsilyl propionate was used as an internal chemical shift and concentration reference. All of the CSF samples showed peaks for lactate, L-alanine, acetate, glutamine, citrate, creatine/creatinine and sugar resonances. There was good agreement between the metabolite concentrations as determined by NMR with those obtained using conventional chemical methods. 1D and 2D 1H NMR techniques along with J-coupling and T1 analysis were used to confirm the peak assignments. Methylmalonic acid could be detected and quantitated (ca 150 microM) in the CSF from the vitamin B12 deficient patient.
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-(+)-Lactic acid solution, 30% in H2O (by weight)
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
L-(+)-Lactic acid solution, in H2O, ≥85%
Sigma-Aldrich
L-(+)-Lactic acid solution, meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
L-(+)-Lactic acid solution, ~40% in H2O
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Citric acid, SAJ first grade, ≥99.5%
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-(+)-Lactic acid, ≥98%
Sigma-Aldrich
L-(+)-Lactic acid, BioXtra, ≥98% (titration)
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)