Merck
Binding of acetaldehyde to human and Guinea pig placentae in vitro.
Placenta (2003-02-05)
Marjie L Hard, Umar Iqbal, James F Brien, Gideon Koren
ABSTRACT
Significant interindividual variability exists following maternal alcohol consumption; not all children born to alcoholic women manifest the symptoms associated with foetal alcohol spectrum disorder (FASD). To investigate the potential role of the placenta as a source of variability by determining if interindividual variability exists in the binding of acetaldehyde to human placenta. Acetaldehyde was added to ten different human placental homogenates and subjected to equilibrium dialysis. Homogenates of placentae obtained from guinea pigs chronically exposed to ethanol throughout gestation were also dialysed in the presence of acetaldehyde to look for alterations in binding after chronic alcohol exposure. Nonlinear least-squares regression analysis was used to characterize the binding system involved. It was found that the amount of acetaldehyde bound to human placentae varied by as much as 3-fold among placentae. The binding profile of acetaldehyde was characterized as a two site binding system (Ka(1)=9.8 x 10(5)+/-0.7 x 10(5)l/mol, N(1)=1.1 x 10(-8)+/-0.7 x 10(-8)mol/g tissue; Ka(2)=1.6 x 10(4)+/-0.9 x 10(4)l/mol, N(2)=1.7 x 10(-7)+/-0.4 x 10(-7)mol/g tissue). Chronic alcohol exposure had no effect on the degree of acetaldehyde binding. This previously unidentified source of variability may partially explain why some foetuses are adversely affected by prenatal alcohol exposure while others are not.
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, ≥99%, meets FCC analytical specification
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, natural, FG