Merck
Combination azelaic acid therapy for acne vulgaris.
Journal of the American Academy of Dermatology (2000-07-18)
G Webster
ABSTRACT
There is no topical antiacne medication that acts against all four of the major pathophysiologic features of acne: hyperkeratinization, sebum production, bacterial proliferation, and inflammation. Topical azelaic acid cream helps both to normalize keratinization and to reduce the proliferation of Propionibacterium acnes, and has proven to be effective against both noninflammatory and inflammatory lesions. The results of a recent study now demonstrate that its efficacy can be enhanced, and patient ratings of overall impression improved, when it is used in combination with other topical medications such as benzoyl peroxide 4% gel, clindamycin 1% gel, tretinoin 0.025% cream, and erythromycin 3%/benzoyl peroxide 5% gel. Furthermore, another study has shown that azelaic acid plus benzoyl peroxide achieves greater efficacy and higher patient ratings of convenience than monotherapy with erythromycin-benzoyl peroxide gel.
Product Description
Supelco
Azelaic acid, analytical standard
Sigma-Aldrich
Azelaic acid, technical, ~85% (GC)
Sigma-Aldrich
Azelaic acid, technical grade, 80%
Sigma-Aldrich
Azelaic acid, 98%