Merck
New paradigms for treatment-resistant depression.
Annals of the New York Academy of Sciences (2013-07-24)
Carlos Zarate, Ronald S Duman, Guosong Liu, Simone Sartori, Jorge Quiroz, Harald Murck
ABSTRACT
Clinical depression is a serious mental disorder characterized by low mood, anhedonia, loss of interest in daily activities, and other symptoms, and is associated with severe consequences including suicide and increased risk of cardiovascular events. Depression affects nearly 15% of the population. The standard of care for the last 50 years has focused on monoamine neurotransmitters, including such treatments as selective serotonin reuptake inhibitors (SSRIs) and serotonin-norepinephrine reuptake inhibitors (SNRIs). However, these treatments have significant limitations: they can take weeks before showing mood-altering effects, and only one to two out of ten patients shows clinical effects beyond those associated with placebo. A major paradigm shift in research into the treatment of depression is underway, based on promising results with the glutamatergic NMDA receptor antagonist ketamine. Further research has demonstrated the significance of glutamatergic pathways in depression and the association of this system with the stress pathway and magnesium homeostasis. Treatment with NMDA receptor antagonists and magnesium have shown the ability to sprout new synaptic connections and reverse stress-induced neural changes, opening up promising new territory for the development of drugs to meet the unmet need in patients with clinical depression.
Sigma-Aldrich
Magnesium, ReagentPlus®, powder, −50 mesh, ≥99%
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, ReagentPlus®, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
(±)-Ketamine hydrochloride, solid
Supelco
Ketamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®