Merck
  • Evidence of AlOHC responsible for the radiation-induced darkening in Yb doped fiber.
Evidence of AlOHC responsible for the radiation-induced darkening in Yb doped fiber.
Optics express (2013-04-11)
Thierry Deschamps, Hervé Vezin, Cédric Gonnet, Nadège Ollier
ABSTRACT
Using a combination of experimental techniques such as optical absorption, Raman scattering, continuous wave and pulse Electron Spin Resonance (ESR), we characterize a set of γ-irradiated Yb(3+) doped silica glass preforms with different contents of phosphorous and aluminum. We demonstrate that when P is introduced in excess compared to Al, nearly no radiodarkening is induced by γ-rays. On the other hand, when Al>P, a large absorption band is induced by radiation. Thermal annealing experiments reveal the correlation between the decrease of the optical absorption band and the decrease of the Al-Oxygen Hole Center (AlOHC) ESR signal, demonstrating the main role of AlOHC defects in the fiber darkening. HYSCORE (HYperfine Sublevel CORElation) pulse-ESR experiments show a high Al-P nuclear spin coupling when P>Al and no coupling when Al>P. This result suggests that both AlOHC and POHC creation is inhibited by Al-O-P linkages. Confronting our data with previous works, we show that the well-known photodarkening process, meaning losses induced by the IR pump, can also be explained in this framework.
Product Description
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, ≤30 μm, spherical powder, ≥99%
Sigma-Aldrich
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Ytterbium, powder, ≥99.9% trace rare earth metals basis
Sigma-Aldrich
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, CP
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Aluminum, IRMM®, certified reference material, 0.1 mm foil