Merck
  • Efficacy and safety of single-dose fosaprepitant in the prevention of chemotherapy-induced nausea and vomiting in patients receiving high-dose cisplatin: a multicentre, randomised, double-blind, placebo-controlled phase 3 trial.
Efficacy and safety of single-dose fosaprepitant in the prevention of chemotherapy-induced nausea and vomiting in patients receiving high-dose cisplatin: a multicentre, randomised, double-blind, placebo-controlled phase 3 trial.
Annals of oncology : official journal of the European Society for Medical Oncology (2012-11-03)
H Saito, H Yoshizawa, K Yoshimori, N Katakami, N Katsumata, M Kawahara, K Eguchi
ABSTRACT
We evaluated the efficacy and safety of single-dose fosaprepitant in combination with intravenous granisetron and dexamethasone. Patients receiving chemotherapy including cisplatin (≥70 mg/m(2)) were eligible. A total of 347 patients (21% had received cisplatin with vomiting) were enrolled in this trial to receive the fosaprepitant regimen (fosaprepitant 150 mg, intravenous, on day 1 in combination with granisetron, 40 μg/kg, intravenous, on day 1 and dexamethasone, intravenous, on days 1-3) or the control regimen (placebo plus intravenous granisetron and dexamethasone). The primary end point was the percentage of patients who had a complete response (no emesis and no rescue therapy) over the entire treatment course (0-120 h). The percentage of patients with a complete response was significantly higher in the fosaprepitant group than in the control group (64% versus 47%, P = 0.0015). The fosaprepitant regimen was more effective than the control regimen in both the acute (0-24 h postchemotherapy) phase (94% versus 81%, P = 0.0006) and the delayed (24-120 h postchemotherapy) phase (65% versus 49%, P = 0.0025). Single-dose fosaprepitant used in combination with granisetron and dexamethasone was well-tolerated and effective in preventing chemotherapy-induced nausea and vomiting in patients receiving highly emetogenic cancer chemotherapy, including high-dose cisplatin.
Product Description
Granisetron hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Granisetron hydrochloride, ≥98% (HPLC), solid