Merck
  • Internal radiotherapy techniques using radiolanthanide praseodymium-142: a review of production routes, brachytherapy, unsealed source therapy.
Internal radiotherapy techniques using radiolanthanide praseodymium-142: a review of production routes, brachytherapy, unsealed source therapy.
Annals of nuclear medicine (2011-07-02)
Mohamadreza K Bakht, Mahdi Sadeghi
ABSTRACT
Radionuclides of rare earth elements are gaining importance as emerging therapeutic agents in nuclear medicine. β(-)-particle emitter 142Pr [T (1/2) = 19.12 h, E(-)β = 2.162 MeV (96.3%), Eγ = 1575 keV (3.7%)] is one of the praseodymium-141 (100% abundant) radioisotopes. Production routes and therapy aspects of 142Pr will be reviewed in this paper. However, 142Pr produces via 141Pr(n, γ) 142Pr reaction by irradiation in a low-fluence reactor; 142Pr cyclotron produced, could be achievable. 142Pr due to its high β(-)-emission and low specific gamma γ-emission could not only be a therapeutic radionuclide, but also a suitable radionuclide in order for biodistribution studies. Internal radiotherapy using 142Pr can be classified into two sub-categories: (1) unsealed source therapy (UST), (2) brachytherapy. UST via 142Pr-HA and 142Pr-DTPA in order for radiosynovectomy have been proposed. In addition, 142Pr Glass seeds and 142Pr microspheres have been utilized for interstitial brachytherapy of prostate cancer and intraarterial brachytherapy of arteriovenous malformation, respectively.
Product Description
Praseodymium, rod, 50mm, diameter 6.35mm, cast, 99%
Praseodymium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Praseodymium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Praseodymium, foil, not light tested, 25x25mm, thickness 0.015mm, as rolled, 99%
Praseodymium, foil, not light tested, 25x25mm, thickness 0.01mm, as rolled, 99%
Praseodymium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Praseodymium, foil, 25x25mm, thickness 0.25mm, as rolled, 99%
Praseodymium, foil, not light tested, 50x50mm, thickness 0.005mm, as rolled, 99%
Praseodymium, foil, not light tested, 50x50mm, thickness 0.015mm, as rolled, 99%
Praseodymium, foil, not light tested, 50x50mm, thickness 0.01mm, as rolled, 99%
Praseodymium, foil, 50x50mm, thickness 0.25mm, as rolled, 99%
Praseodymium, rod, 100mm, diameter 6.35mm, cast, 99%