Merck
  • [Evaluation of the biological effects of chemical substances on albino rat offspring].
[Evaluation of the biological effects of chemical substances on albino rat offspring].
Gigiena i sanitariia (2012-01-19)
V S Rukavishnikov, L M Sosedova, E A Kapustina
ABSTRACT
The paper provides the results of experimental studies of the impact of vinyl chloride or sublimate intoxication in male albino rats on the functional state of the nervous system of their offspring. Retarded sensorimotor development was revealed in the neonatal offspring. The pubertal offspring showed behavioral pattern disintegrity that was characterized by changes in motor activity and orientative-exploratory responses and by anxiety. There was abnormal impulse conduction in the neuromuscular apparatus of the hind legs of albino rats and morphological changes in the structure of nervous tissue.
Product Description
Supelco
Vinyl chloride solution, certified reference material, 2000 μg/mL in methanol
Supelco
Vinyl chloride solution, 200 μg/mL in methanol, analytical standard