Merck
  • Rapid extraction, radioiodination, and in vivo catabolism of 125I-labeled fibrinogen in the horse.
Rapid extraction, radioiodination, and in vivo catabolism of 125I-labeled fibrinogen in the horse.
American journal of veterinary research (1985-12-01)
C P Coyne, W J Hornof, A B Kelly, T R O'Brien, S J DeNardo
ABSTRACT
Two methods were analyzed for the rapid extraction of equine fibrinogen from fresh plasma, using ammonium sulfate-sodium phosphate buffer. Fibrinogen from each of these 2 methods was then radiolabeled with 125I (half-life = 60.2 days, gamma = 35 keV), using monochloroiodine reagent. Mean protein-bound activity was 98.5% and mean clottable radioactivity was 94.1%. Radiolabeled fibrinogen administered IV to 15 horses had an overall mean (+/- SD) plasma half-life of 4.95 +/- 0.44 days.
Sigma-Aldrich
Iodine monochloride, 99.998% trace metals basis
Sigma-Aldrich
Wijs (iodine monochloride) solution | Chloroiodide | 7790-99-0 solution
Sigma-Aldrich
Iodine monochloride solution, 1 M in acetic acid
Sigma-Aldrich
Iodine monochloride, reagent grade, ≥95%
Sigma-Aldrich
Iodine monochloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Iodine monochloride, ACS reagent, 1.10±0.1 I/Cl ratio basis