Merck
  • Brand of dipotassium EDTA vacuum tube as a new source of pre-analytical variability in routine haematology testing.
Brand of dipotassium EDTA vacuum tube as a new source of pre-analytical variability in routine haematology testing.
British journal of biomedical science (2013-04-27)
G Lima-Oliveira, G Lippi, G L Salvagno, M Montagnana, G Poli, G P Solero, G Picheth, G C Guidi
ABSTRACT
This study assesses the use of different dry K2 (dipotassium) EDTA vacuum tubes and whether or not they might represent a bias in haematological testing. Blood was collected in three dipotassium EDTA vacuum tubes from different manufacturers: Venosafe, Vacuette and Vacutainer. Samples were analysed on an Advia 2120i analyser. Significant differences among results and biases were compared with current quality specifications. Significant differences were found for haematocrit (HCT), mean corpuscular volume (MCV), white blood cell count (WBC) and platelet distribution width (PDW) when comparing Venosafe vs. Vacuette; for MCV, WBC and PDW when comparing Venosafe vs. Vacutainer; and for HCT and MCV when comparing Vacuette vs. Vacutainer. Clinically significant variations were observed for HCT and PDW in Venosafe vs. Vacuette; PDW in Venosafe vs. Vacutainer; and HCT and MCV in Vacuette vs. Vacutainer. The use of dipotassium EDTA vacuum tubes from different manufacturers represent a clinically relevant source of variation for HCT, MCV and PDW.
Product Description
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 0.1 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 0.01 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 0.02 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 0.05 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution