Merck
  • A simple iterative method for the synthesis of β-(1→6)-glucosamine oligosaccharides.
A simple iterative method for the synthesis of β-(1→6)-glucosamine oligosaccharides.
Carbohydrate research (2013-03-26)
Lucy G Weaver, Yogendra Singh, Joanne T Blanchfield, Paul L Burn
ABSTRACT
Poly-N-acetylglucosamine (PNAG) saccharides are an important constituent of bacterial biofilms, such as those produced by Staphylococcus aureus. We have developed a simple two-step iterative method for the synthesis of β-(1→6)-glucosamine oligosaccharides that are structurally similar to PNAG. We illustrate the method with the formation of a pentasaccharide. The key building block is an orthogonally protected N-trifluoroacetamido thioglycoside donor that was added in succession to a glycosyl acceptor, enabling efficient glycosylation of the growing chain. In the second step of the iterative cycle, this building block is quantitatively deprotected at the C-6-hydroxyl position, ready for the next saccharide addition. Building from an azido-functionalised GlcNAc monosaccharide acceptor, the pentasaccharide was synthesised in seven steps in an overall yield of 25%.
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥95% (HPLC)
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥99% (HPLC)
Sigma-Aldrich
N-Acetyl-D-glucosamine, BioReagent, suitable for cell culture