Merck
  • Dielectric behavior of gelatine-glycosaminoglycans blends: an impedance analysis.
Dielectric behavior of gelatine-glycosaminoglycans blends: an impedance analysis.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Ivy Kanungo, Nishter Nishad Fathima, Jonnalagadda Raghava Rao
ABSTRACT
The dielectric behavior of the gelatine-GAGs based blend systems has been studied to understand the dynamic behavior of the water at the protein-GAGs interfaces which are relevant for tissue engineering application. Impedance (Z) and phase have been measured as a function of frequencies from 0.01 Hz to 100 kHz. GAGs tunes the ionic charge drift which initiates polarization mechanisms through charge accumulation at structural interfaces and creates conduction currents. The admittance results showed that at high frequency, the conductivity increases with increasing GAGs concentration indicating changes in hydration shell of the gelatine by the GAGs.
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Genipin, ≥98% (HPLC), powder
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength