Merck
  • Total, elastic, and inelastic cross sections for positron and electron collisions with tetrahydrofuran.
Total, elastic, and inelastic cross sections for positron and electron collisions with tetrahydrofuran.
The Journal of chemical physics (2013-03-01)
Luca Chiari, Emma Anderson, Wade Tattersall, J R Machacek, Prasanga Palihawadana, Casten Makochekanwa, James P Sullivan, Gustavo García, Francisco Blanco, R P McEachran, M J Brunger, Stephen J Buckman
ABSTRACT
We present total, elastic, and inelastic cross sections for positron and electron scattering from tetrahydrofuran (THF) in the energy range between 1 and 5000 eV. Total cross sections (TCS), positronium formation cross sections, the summed inelastic integral cross sections (ICS) for electronic excitations and direct ionization, as well as elastic differential cross sections (DCS) at selected incident energies, have been measured for positron collisions with THF. The positron beam used to carry out these experiments had an energy resolution in the range 40-100 meV (full-width at half-maximum). We also present TCS results for positron and electron scattering from THF computed within the independent atom model using the screening corrected additivity rule approach. In addition, we calculated positron-impact elastic DCS and the sum over all inelastic ICS (except rotations and vibrations). While our integral and differential positron cross sections are the first of their kind, we compare our TCS with previous literature values for this species. We also provide a comparison between positron and electron-impact cross sections, in order to uncover any differences or similarities in the scattering dynamics with these two different projectiles.
Product Description
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%