Merck
  • Visible-light-mediated thiol-ene hydrogelation using eosin-Y as the only photoinitiator.
Visible-light-mediated thiol-ene hydrogelation using eosin-Y as the only photoinitiator.
Macromolecular rapid communications (2013-02-07)
Han Shih, Chien-Chi Lin
ABSTRACT
The utility of visible-light-mediated polymerization in tissue engineering has been limited due to the necessary use of potentially cytotoxic coinitiator and comonomer. Here, we report a visible-light-mediated thiol-ene hydrogelation scheme using eosin-Y as the only photoinitiator. Under visible light exposure, rapid and highly tunable step-growth gelation is achieved using PEG-norbornene and a model cross-linker dithiothreitol. In addition to investigating the gelation kinetics and properties of thiol-ene hydrogels formed by this new gelation scheme, we also report high cytocompatibility of these hydrogels using human mesenchymal stem cells (hMSCs) and pancreatic MIN6 β-cells.
Sigma-Aldrich
Eosin Y, SAJ special grade, ≥85.0%
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y disodium salt, certified by the Biological Stain Commission, certified by the BSC
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O