Merck
  • Magnetic loading of graphene-nickel nanoparticle hybrid for electrochemical sensing of carbohydrates.
Magnetic loading of graphene-nickel nanoparticle hybrid for electrochemical sensing of carbohydrates.
Biosensors & bioelectronics (2012-12-19)
Weidong Qu, Luyan Zhang, Gang Chen
ABSTRACT
Graphene-nickel nanoparticle hybrid was prepared by the one-step far infrared-assisted reduction of graphene oxide and nickel (II) ions using hydrazine. It was loaded on the surface of a magnetic electrode for electrochemical sensing. The feasibility and performance of the novel electrode were demonstrated by measuring carbohydrates using cyclic voltammetry and amperometry. It demonstrated that nickel nanoparticles decorated on graphene sheets exhibited higher electrocatalytic activity toward the oxidation of carbohydrates while graphene improved the electron transduction. The synergistic effect significantly enhanced the current response of carbohydrates.
Sigma-Aldrich
Hydrazine monohydrate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Hydrazine solution, 1 M in acetonitrile
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazine solution, 1.0 M in THF
Sigma-Aldrich
Hydrazine solution, 35 wt. % in H2O
Sigma-Aldrich
Hydrazine hemisulfate salt, ≥98%
Sigma-Aldrich
Hydrazinium dichloride, SAJ special grade, ≥98.0%
Sigma-Aldrich
Hydrazine solution, 1.0 M in ethanol
Sigma-Aldrich
Hydrazine sulfate salt, puriss. p.a., ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrazine dihydrochloride, ≥98%
Sigma-Aldrich
Hydrazine sulfate salt, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrazine, anhydrous, 98%