Merck
  • Glycopolymer micelles with reducible ionic cores for hepatocytes-targeting delivery of DOX.
Glycopolymer micelles with reducible ionic cores for hepatocytes-targeting delivery of DOX.
International journal of pharmaceutics (2012-12-15)
Yanxia Wang, Xinge Zhang, Peien Yu, Chaoxing Li
ABSTRACT
A novel galactose-decorated cross-linked micelles (cl-micelles) with ionic cores using cystamine (Cys) as a biodegradable cross-linker was prepared by using block ionomer complexes of poly(ethylene glycol)-b-poly(2-acryloxyethyl-galactose)-b-poly(acrylic acid) (PEG-b-PAEG-b-PAA) and Ca(2+) (PEG-b-PAEG-b-PAA cl-micelles/Cys). Doxorubicin (DOX) was successfully incorporated into the ionic cores of such micelles via electrostatic interactions. Proton nuclear magnetic resonance spectrum and Fourier transform infrared spectrometer indicated galactose ligands were exposed at the micellar surface. The micelles were spherical in shape, with an average size of 100nm. The in vitro release studies confirmed that DOX-loaded PEG-b-PAEG-b-PAA cl-micelles/Cys accomplished rapid drug release under reducing condition. Remarkably, PEG-b-PAEG-b-PAA cl-micelles/Cys efficiently delivered and released DOX into the cell nucleus of HepG2 cells, and the intensity of fluorescence observed in HepG2 cells was stronger than that incubated with the micelles without galactose ligands. In contrast, little fluorescence was observed in NIH3T3 cells after incubation with PEG-b-PAEG-b-PAA cl-micelles/Cys. Interestingly, cytotoxicity assays showed that DOX-loaded PEG-b-PAEG-b-PAA cl-micelles/Cys retained higher cell inhibition efficiency in HepG2 cells as compared with NIH3T3 cells, and were more potent than the micelles without galactose ligands and the micelles with non degradable cross-links. These results indicate that PEG-b-PAEG-b-PAA cl-micelles/Cys have great potential in liver tumor-targeted chemotherapy.
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~2,000, 50 wt. % in H2O, electronic grade
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~250,000, 35 wt. % in H2O
Supelco
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)
Sigma-Aldrich
Poly(acrylic acid), average Mv ~1,250,000
Sigma-Aldrich
Poly(acrylic acid)
Sigma-Aldrich
Cystamine dihydrochloride, BioXtra
Sigma-Aldrich
Cystamine dihydrochloride, 96%
Sigma-Aldrich
Cystamine dihydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Poly(acrylic acid), average Mv ~3,000,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~4,000,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~450,000