Merck
  • Self-assembled magnetic theranostic nanoparticles for highly sensitive MRI of minicircle DNA delivery.
Self-assembled magnetic theranostic nanoparticles for highly sensitive MRI of minicircle DNA delivery.
Nanoscale (2012-12-12)
Qian Wan, Lisi Xie, Lin Gao, Zhiyong Wang, Xiang Nan, Hulong Lei, Xiaojing Long, Zhi-Ying Chen, Cheng-Yi He, Gang Liu, Xin Liu, Bensheng Qiu
ABSTRACT
As a versatile gene vector, minicircle DNA (mcDNA) has a great potential for gene therapy. However, some serious challenges remain, such as to effectively deliver mcDNA into targeted cells/tissues and to non-invasively monitor the delivery of the mcDNA. Superparamagnetic iron oxide (SPIO) nanoparticles have been extensively used for both drug/gene delivery and diagnosis. In this study, an MRI visible gene delivery system was developed with a core of SPIO nanocrystals and a shell of biodegradable stearic acid-modified low molecular weight polyethyleneimine (Stearic-LWPEI) via self-assembly. The Stearic-LWPEI-SPIO nanoparticles possess a controlled clustering structure, narrow size distribution and ultrasensitive imaging capacity. Furthermore, the nanoparticle can effectively bind with mcDNA and protect it from enzymatic degradation. In conclusion, the nanoparticle shows synergistic advantages in the effective transfection of mcDNA and non-invasive MRI of gene delivery.
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Stearic acid, SAJ first grade, ≥90.0%, powder
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Supelco
Stearic acid, analytical standard