Merck
  • Pharmacokinetics of polymyxin B in patients on continuous venovenous haemodialysis.
Pharmacokinetics of polymyxin B in patients on continuous venovenous haemodialysis.
The Journal of antimicrobial chemotherapy (2012-11-28)
Ana M Sandri, Cornelia B Landersdorfer, Jovan Jacob, Márcio M Boniatti, Micheline G Dalarosa, Diego R Falci, Tainá F Behle, David Saitovitch, Jiping Wang, Alan Forrest, Roger L Nation, Alexandre P Zavascki, Jian Li
ABSTRACT
To evaluate the pharmacokinetics of polymyxin B in patients on continuous venovenous haemodialysis (CVVHD) after intravenous administration of unadjusted dosage regimens. Two critically ill patients had eight blood samples collected during a 12 h interval on days 8 and 10 of polymyxin B therapy. Dialysate was collected every hour during the 12 h dosing interval. Polymyxin B binding in plasma was determined by rapid equilibrium dialysis. Concentrations of polymyxin B in plasma and dialysate samples were quantified using a validated ultra-performance liquid chromatography-tandem mass spectrometry assay. Respective maximum plasma concentrations in patients 1 and 2 were 8.62 and 4.38 mg/L; total body clearances (scaled linearly by body weight) were 0.043 and 0.027 L/h/kg, respectively, of which 12.2% and 5.62% were dialysis clearance, respectively. The corresponding volumes of distribution of polymyxin B at steady state were 0.50 and 0.34 L/kg, respectively, and protein binding in pooled plasma samples was 74.1% and 48.8%, respectively. Our findings indicate that the recommended polymyxin B doses should not be reduced for patients on CVVHD.
Sigma-Aldrich
Polymyxin B solution, 20 mg/mL in H2O
Supelco
Polymyxin B solution, 1 mg/mL in H2O, analytical standard
Sigma-Aldrich
Polymyxin B sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Polymyxin B sulfate salt
Sigma-Aldrich
Polymyxin B sulfate, meets USP testing specifications
Polymyxin B sulfate, European Pharmacopoeia (EP) Reference Standard