Merck
  • Immune responses and gene expression in hepatopancreas from Macrobrachium rosenbergii challenged by a novel pathogen spiroplasma MR-1008.
Immune responses and gene expression in hepatopancreas from Macrobrachium rosenbergii challenged by a novel pathogen spiroplasma MR-1008.
Fish & shellfish immunology (2012-11-28)
Jie Du, Huanxi Zhu, Peng Liu, Jing Chen, Yunji Xiu, Wei Yao, Ting Wu, Qian Ren, Qingguo Meng, Wei Gu, Wen Wang
ABSTRACT
Freshwater prawn Macrobrachium rosenbergii inoculated with 100 μl novel pathogen spiroplasma strain MR-1008 in logarithmic phase (10(8) spiroplasmas ml(-1)) were examined for alkaline phosphatase (AKP) activity, acid phosphatase (ACP) activity, superoxide dismutase (SOD) activity, catalase (CAT) activity, as well as expressions of 7 immune related genes in hepatopancreas after 1-28 d. Hematoxylin-eosin (HE) staining showed obvious pathological features in hepatopancreas connective and epithelial tissue. Enzyme activity analyze showed that hepatopancreas AKP and ACP activity increased markedly (P < 0.05) when inoculated with spiroplasma MR-1008 after 5 d and 10 d, respectively. SOD enzyme activity changed less obviously and slightly increased at 1 day post-inoculation, but CAT activity decreased significantly after 5 d inoculation. The expression levels of lipopolysaccharide and β-1,3-glucan-binding protein (LGBP), peroxinectin (PE), α2-macroglobulin (α2M), AKP, ACP, CAT, and copper/zinc SOD (Cu, Zn-SOD) genes in the hepatopancreas were examined by Real-Time PCR (qRT-PCR) and the results demonstrated that these immune related genes were induced by challenge with spiroplasma MR-1008. The results suggested that the prawn immune responses could be activated or inhibited by spiroplasma MR-1008, and that the hepatopancreas also plays key roles in innate immunity for defense against the pathogen.
Sigma-Aldrich
Eosin Y, SAJ special grade, ≥85.0%
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Eosin Y disodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Hematoxylin monohydrate, SAJ special grade
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission