Merck
Total synthesis of alotaketal A.
Organic letters (2012-10-26)
Mengyang Xuan, Ian Paterson, Stephen M Dalby
ABSTRACT
The total synthesis of the cAMP signaling pathway activator (-)-alotaketal A is reported. A convergent approach to the unusual alotane sesterterpenoid skeleton was employed, exploiting a remarkable LiDBB-mediated coupling of an (R)-carvone-derived δ-lactone with an allyl bromide side chain, followed by spiroacetalization.
Sigma-Aldrich
L-Carvone, natural, 99%, FG
Sigma-Aldrich
L-Carvone, ≥97%, FCC, FG
Supelco
(+)-Carvone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
(R)-(−)-Carvone, 98%
Supelco
(−)-Carvone, analytical standard
Sigma-Aldrich
D-Carvone, ≥96%, FG
Supelco
(+)-Carvone, analytical standard
Sigma-Aldrich
(S)-(+)-Carvone, 96%