Merck
  • Purification and characterization of carbonic anhydrase from sheep kidney and effects of sulfonamides on enzyme activity.
Purification and characterization of carbonic anhydrase from sheep kidney and effects of sulfonamides on enzyme activity.
Bioorganic & medicinal chemistry (2012-09-15)
Ramazan Demirdağ, Veysel Çomaklı, Murat Şentürk, Deniz Ekinci, Ö İrfan Küfrevioğlu, Claudiu T Supuran
ABSTRACT
Carbonic anhydrase (CA, EC: 4.2.1.1) was purified from sheep kidney by affinity chromatography on a Sepharose 4B-tyrosine-sulfanilamide column. By means of two consecutive procedures, the enzyme (sCA) was purified 227.61-fold with a yield of 60.75%, and a specific activity of 838.89U/mg proteins. The optimum temperature, ionic strength and pH were determined to be 35°C, 20mM and 8.5, respectively. The molecular weight determined by SDS-PAGE was found to be 29kDa. The kinetic parameters, KM and Vmax values were determined for the 4-nitrophenyl acetate (p-NpA) hydrolysis reaction. Some sulfonamides were tested as inhibitors against the purified CAs enzyme. The Ki constants for benzenesulfonamide (1), sulfanilamide (2), mafenide (3), 4-(2-aminoethyl) benzenesulfonamide (4), 4-methyl-benzenesulfonamide (5), 2-bromo-benzenesulfonamide (6), naphthalene-2-sulfonamide (7), 4-amino-6-chlorobenzene-1,3-disulfonamide (8) and saccharin (9) were in the range 1.348-69.31μM.
Product Description
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, ≥98%
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)