Merck
  • Enhancing the power of liquid chromatography-mass spectrometry-based urine metabolomics in negative ion mode by optimization of the additive.
Enhancing the power of liquid chromatography-mass spectrometry-based urine metabolomics in negative ion mode by optimization of the additive.
Analytical chemistry (2012-08-15)
Xumin Zhang, Morten Rahr Clausen, Xiaolu Zhao, Hong Zheng, Hanne Christine Bertram
ABSTRACT
Untargeted liquid chromatography-mass spectrometry (LC-MS)-based metabolomics studies are usually carried out in both positive and negative ion modes; however, it is frequently ignored that the optimal conditions in positive ion mode and negative ion mode are often not the same. We carried out a systematic investigation on urine samples to evaluate the additive effects in negative ion mode. It was found that the widely used conditions, 0.1% formic acid (FA) and NH(4)Ac at different pH, are far from the optimum for untargeted urine metabolomics studies. Compared to 0.1% FA, the use of 1 mM acetic acid (HAc) resulted in almost three times as many detected peaks (401 vs 148) and around five times the size of the peak area (33.55 × 10(6) vs 6.47 × 10(6)). The remarkable improvement can be explained by two factors: (i) a significantly enhanced ionization efficiency due to the combination of an appropriate pH at around 4.0-4.5, the reducibility of H(+), and the high gas-phase basicity of Ac(-) and (ii) a reproducible LC separation due to an acceptable buffering capacity. Our study revealed the importance and necessity of additive optimization, which can be of benefit in related metabolomics studies.
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v), for copper determination
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis