Merck
  • Analysis of alkylresorcinols in cereal grains and products using ultrahigh-pressure liquid chromatography with fluorescence, ultraviolet, and CoulArray electrochemical detection.
Analysis of alkylresorcinols in cereal grains and products using ultrahigh-pressure liquid chromatography with fluorescence, ultraviolet, and CoulArray electrochemical detection.
Journal of agricultural and food chemistry (2012-08-14)
Alastair B Ross
ABSTRACT
Alkylresorcinols are phenolic lipids, with homologues ranging from C17 to C25, found in high concentrations in whole grain wheat and rye, lower concentrations in barley, and negligible concentrations in refined wheat flour. The analysis of alkylresorcinols is of importance due to their potential as biomarkers of whole grain intake and emerging evidence for some biological effects. Present HPLC methods have insufficient resolution for accurately quantitating the mix of alkyl- and alkenylresorcinols found in rye. An ultrahigh-pressure liquid chromatography method was developed, and three detection methods (CoulArray (CAED), ultraviolet (UV), and fluorescence detection (FD)) were compared for cereal alkylresorcinol analysis. The lower limits of quantitation and detection were 50 and 20 pg injected, 5 pg and 2 pg injected, and 500 and 1250 pg injected for FD, CAED, and UV, respectively. FD and CAED provided similar results, with some bias for higher results with FD (<10% difference). UV detection generally resulted in overestimation of alkylresorcinol concentrations. The method was applied to cereal (15) and cereal product (90) samples mainly from the United States with results in the same range as previous methods. The improved resolution with this method allows facile analysis of alkylresorcinols from cereal products, including minor unsaturated homologues such as those found in rye.
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Resorcinol, JIS special grade, ≥99.0%
Sigma-Aldrich
Resorcinol, SAJ first grade, ≥98.0%
Supelco
Resorcinol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%