Merck
  • Charge-transfer guest interactions in luminescent MOFs: implications for solid-state temperature and environmental sensing.
Charge-transfer guest interactions in luminescent MOFs: implications for solid-state temperature and environmental sensing.
Dalton transactions (Cambridge, England : 2003) (2012-06-19)
Patrick L Feng, Kirsty Leong, Mark D Allendorf
ABSTRACT
One of the ongoing goals in the field of porous materials is the design and synthesis of materials that possess chemical structures amenable for use in sensing applications. We describe the preparation, luminescence characteristics, and environmental sensing properties of variants of the aluminum-based MOF [Al(OH)(O(2)C-C(10)H(6)-CO(2))](∞). Careful activation of the open framework complex, 1, yielded a dynamic structural transformation to a non-porous form, 2, that exhibited strong inter-linker interactions and red-shifted emission characteristics indicative of dimer formation. We also demonstrate the formation of highly luminescent ground-state charge-transfer (CT) complexes between 1 and the electron-donating amines dimethylamine (DMA) (1a), and N,N-diethylaniline (DEA) (1b), both of which exhibit dual-emission characteristics and a ratiometric luminescence response that is sensitive to temperature and solvent polarity. Steady-state and time-resolved measurements on 1a, 1b, and 2 indicate that the MOF structures stabilize ground-state CT interactions that are distinct from the weakly-bound exciplexes formed in comparable mixtures of purely organic components. The spectra for 1a and 1b also indicate different temperature dependencies that correspond to thermally-activated complex formation (ΔH(f) = +1.1 ± 0.2 kcal mol(-1)) in 1a and static quenching effects (ΔH(f) = -2.2 ± 0.3 kcal mol(-1)) in 1b. The addition of ethanol, isopropanol, toluene, or chloroform to suspensions of 1b indicate destabilization of the CT state with increasing solvent polarity, which suggests the generalized application of this or related materials in sensor applications.
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol
Sigma-Aldrich
Dimethylamine hydrochloride, 99%
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF
Sigma-Aldrich
Dimethylamine hydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Dimethylamine solution, CP, 50% in H2O
Sigma-Aldrich
Dimethylamine solution, 40 wt. % in H2O