Merck
  • Enzymatic mineralization of hydrogels for bone tissue engineering by incorporation of alkaline phosphatase.
Enzymatic mineralization of hydrogels for bone tissue engineering by incorporation of alkaline phosphatase.
Macromolecular bioscience (2012-06-01)
Timothy E L Douglas, Philip B Messersmith, Safak Chasan, Antonios G Mikos, Eric L W de Mulder, Glenn Dickson, David Schaubroeck, Lieve Balcaen, Frank Vanhaecke, Peter Dubruel, John A Jansen, Sander C G Leeuwenburgh
ABSTRACT
Alkaline phosphatase (ALP), an enzyme involved in mineralization of bone, is incorporated into three hydrogel biomaterials to induce their mineralization with calcium phosphate (CaP). These are collagen type I, a mussel-protein-inspired adhesive consisting of PEG substituted with catechol groups, cPEG, and the PEG/fumaric acid copolymer OPF. After incubation in Ca-GP solution, FTIR, EDS, SEM, XRD, SAED, ICP-OES, and von Kossa staining confirm CaP formation. The amount of mineral formed decreases in the order cPEG > collagen > OPF. The mineral:polymer ratio decreases in the order collagen > cPEG > OPF. Mineralization increases Young's modulus, most profoundly for cPEG. Such enzymatically mineralized hydrogel/CaP composites may find application as bone regeneration materials.
Sigma-Aldrich
Fumaric acid, FCC, FG
Sigma-Aldrich
Minimum Essential Medium Eagle, 10 ×, With Earle′s salts, without L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fumaric acid, ≥99.0% (T)
Supelco
Fumaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Fumaric acid, European Pharmacopoeia (EP) Reference Standard