Merck
  • The native serotonin 5-HT(5A) receptor: electrophysiological characterization in rodent cortex and 5-HT(1A)-mediated compensatory plasticity in the knock-out mouse.
The native serotonin 5-HT(5A) receptor: electrophysiological characterization in rodent cortex and 5-HT(1A)-mediated compensatory plasticity in the knock-out mouse.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2012-04-28)
Nathalie M Goodfellow, Craig D C Bailey, Evelyn K Lambe
ABSTRACT
The 5-HT(5A) receptor is the least understood serotonin (5-HT) receptor. Here, we electrophysiologically identify and characterize a native 5-HT(5A) receptor current in acute ex vivo brain slices of adult rodent prefrontal cortex. In the presence of antagonists for the previously characterized 5-HT(1A) and 5-HT₂ receptors, a proportion of layer V pyramidal neurons continue to show 5-HT-elicited outward currents in both rats and mice. These 5-HT currents are suppressed by the selective 5-HT(5A) antagonist, SB-699551, and are not observed in 5-HT(5A) receptor knock-out mice. Further characterization reveals that the 5-HT(5A) current is activated by submicromolar concentrations of 5-HT, is inwardly rectifying with a reversal potential near the equilibrium potential for K+ ions, and is suppressed by blockers of Kir3 channels. Finally, we observe that genetic deletion of the inhibitory 5-HT(5A) receptor results in an unexpected, large increase in the inhibitory 5-HT(1A) receptor currents. The presence of functional prefrontal 5-HT(5A) receptors in normal rodents along with compensatory plasticity in 5-HT(5A) receptor knock-out mice testifies to the significance of this receptor in the healthy prefrontal cortex.
Sigma-Aldrich
Barium chloride dihydrate, ≥99.999% trace metals basis
Sigma-Aldrich
Barium chloride dihydrate, ≥99%
Sigma-Aldrich
Barium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.95% trace metals basis
Sigma-Aldrich
Barium chloride, 99.999% trace metals basis
Sigma-Aldrich
Barium chloride, 99.9% trace metals basis
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Anti-Serotonin Receptor 1A Antibody, serum, Chemicon®