Merck
  • Bicyclo[4.4.0]decane oxygenated sesquiterpenes from Eryngium maritimum essential oil.
Bicyclo[4.4.0]decane oxygenated sesquiterpenes from Eryngium maritimum essential oil.
Planta medica (2012-01-20)
Florent Darriet, Mourad Bendahou, Jean-Marie Desjobert, Jean Costa, Alain Muselli
ABSTRACT
Investigation of the essential oil of the aerial parts of Eryngium maritimum L. from Corsica led to the isolation of one known sesquiterpene (1) and three new oxygenated sesquiterpenes with a muurolane or cadinane skeleton (2-4). Structure assignments of 4 βH -muurol-9-en-15-al (1), 4 βH -cadin-9-en-15-al (2), 4 βH -muurol-9-en-15-ol (3) and 4 βH -cadin-9-en-15-ol (4) were carried out by GC/MS (EI and CI) and comprehensive 1D and 2D NMR spectroscopy. Antibacterial activity of Eryngium maritimum L. oil and column chromatography fractions have been investigated for the first time. The oxygenated fraction, which contains the four sesquiterpenes, was efficient against Listeria monocytogenes and Echerichia coli.
Sigma-Aldrich
Decane, SAJ special grade, ≥99.0%
Supelco
Decane, analytical standard
Sigma-Aldrich
Decane, ≥95%
Sigma-Aldrich
Decane, anhydrous, ≥99%
Sigma-Aldrich
Decane, ReagentPlus®, ≥99%
Supelco
Decane, analytical standard