Merck
  • Effect of pluronic P123 and F127 block copolymer on P-glycoprotein transport and CYP3A metabolism.
Effect of pluronic P123 and F127 block copolymer on P-glycoprotein transport and CYP3A metabolism.
Archives of pharmacal research (2011-11-15)
Yanbin Guan, Jiangeng Huang, Lan Zuo, Jiaqiang Xu, Luqin Si, Jun Qiu, Gao Li
ABSTRACT
The aim of the present study was to evaluate the effect of pluronic P123 (P123) and pluronic F127 (F127) on intestinal P-glycoprotein (P-gp) and cytochrome P450 3A using the specific substrates rhodamine-123 (R-123) and midazolam, respectively. Caco-2 cells and everted gut sacs were used as models of intestinal mucosa to assess intestinal absorption of R-123, while rat intestinal microsomes were utilized to examine the effect of P123 and F127 on in vitro midazolam metabolism. P123 and F127 were observed to increase the intracellular accumulation of R-123 in Caco-2 cells in a dose-dependent manner. P123 significantly lowered the efflux ratio of R-123 at two concentrations in Caco-2 monolayers, whereas F127 lowered the efflux ratio only at 1%. Moreover, both pluronics markedly enhanced mucosal to serosal absorption of R-123 in excised ileum of rats. However, no significant difference in relative enzyme activity were observed between P123- or F127-treated and control groups, regardless of the concentrations of P123 and F127 studied. Collectively, these results obtained from the present study demonstrated that P123 and F127 were capable of inhibiting the intestinal P-gp activity, but had little or no effect on intestinal cytochrome P450 3A activity, indicating that P123 and F127 can potentially be used as pharmaceutical ingredients to improve the oral bioavailability of coadministered P-gp substrates via P-gp efflux pump inhibition.
Sigma-Aldrich
Poloxamer 188, micronized
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000
Sigma-Aldrich
Poloxamer 407, micronized
Sigma-Aldrich
Synperonic® PE/P84
Sigma-Aldrich
Synperonic® PE P105, surfactant
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~3,300
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poloxamer 188, solid
Sigma-Aldrich
Poloxamer 407, purified, non-ionic
Sigma-Aldrich
Synperonic® F 108, surfactant, non-ionic
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
Sigma-Aldrich
Poloxamer 188
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800
Sigma-Aldrich
Poloxamer 407, oxyethylene 71.5-74.9 %