Merck
  • Deuterium-isotope study on the reductive ring opening of benzylidene acetals.
Deuterium-isotope study on the reductive ring opening of benzylidene acetals.
Organic & biomolecular chemistry (2011-09-17)
I-Chi Lee, Medel Manuel L Zulueta, Chi-Rung Shie, Susan D Arco, Shang-Cheng Hung
ABSTRACT
Specific deuterated reference compounds were prepared to probe the stereoselectivity of the reductive ring opening of carbohydrate-based benzylidene-type acetals. AlD(3) revealed a retentive stereoselectivity probably through the rare S(N)i (internal nucleophilic substitution) mechanism. An S(N)1-like mechanism occurs in the acid-promoted regioselective BD(3)·THF- or Et(3)SiD-reductive ring opening.
Sigma-Aldrich
Lithium aluminum hydride solution, 2.3 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium aluminum hydride, hydrogen-storage grade
Sigma-Aldrich
Lithium aluminum hydride, pellets, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Lithium aluminum hydride, powder, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric)
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride solution, 2.0 M in THF