Merck
Black stain - a review.
Oral health & preventive dentistry (2011-05-20)
Valerie Ronay, Thomas Attin
ABSTRACT
The purpose of this review was to summarise the fundamentals about black stain, its diagnosis and possible differential diagnoses as well as its microbiology and therapy. In addition, various studies investigating the relationship between black stain and dental caries are examined. Many studies report lower caries prevalence in children with black stain, but this finding could not be confirmed by all authors. Also, a negative relation between degree of staining and caries severity has been described. Reasons for these results are not yet clear but it was speculated that they are related to the specific oral microflora described in black stain-affected individuals.
Sigma-Aldrich
Iron(II) sulfide, SAJ first grade, ≥50.0%
Sigma-Aldrich
Iron(II) sulfide, sticks (thin)
Sigma-Aldrich
Iron(II) sulfide, −100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(II) sulfide, technical grade