Merck
  • Low concentrations of nitric oxide delay the differentiation of embryonic stem cells and promote their survival.
Low concentrations of nitric oxide delay the differentiation of embryonic stem cells and promote their survival.
Cell death & disease (2011-03-04)
J R Tejedo, R Tapia-Limonchi, S Mora-Castilla, G M Cahuana, A Hmadcha, F Martin, F J Bedoya, B Soria
ABSTRACT
Nitric oxide (NO) is an intracellular messenger in several cell systems, but its contribution to embryonic stem cell (ESC) biology has not been characterized. Exposure of ESCs to low concentrations (2-20 μM) of the NO donor diethylenetriamine NO adduct confers protection from apoptosis elicited by leukaemia inhibitory factor (LIF) withdrawal. NO blocked caspase 3 activation, PARP degradation, downregulation of the pro-apoptotic genes Casp7, Casp9, Bax and Bak1 and upregulation of the anti-apoptotic genes Bcl-2 111, Bcl-2 and Birc6. These effects were also observed in cells overexpressing eNOS. Exposure of LIF-deprived mESCs to low NO prevented the loss of expression of self-renewal genes (Oct4, Nanog and Sox2) and the SSEA marker. Moreover, NO blocked the differentiation process promoted by the absence of LIF and bFGF in mouse and human ESCs. NO treatment decreased the expression of differentiation markers, such as Brachyury, Gata6 and Gata4. Constitutive overexpression of eNOS in cells exposed to LIF deprivation maintained the expression of self-renewal markers, whereas the differentiation genes were repressed. These effects were reversed by addition of the NOS inhibitor L-NMMA. Altogether, the data suggest that low NO has a role in the regulation of ESC differentiation by delaying the entry into differentiation, arresting the loss of self-renewal markers and promoting cell survival by inhibiting apoptosis.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Parp1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pou5f1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akt1
Sigma-Aldrich
Diethylenetriamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Diethylenetriamine, ReagentPlus®, 99%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nanog