Merck
  • Prevention of erosive/abrasive enamel wear due to orange juice modified with dietary supplements.
Prevention of erosive/abrasive enamel wear due to orange juice modified with dietary supplements.
Oral diseases (2011-02-22)
Fj Wegehaupt, N Günthart, B Sener, T Attin
ABSTRACT
The aim of this study was to evaluate the erosive/abrasive enamel wear after contact with orange juices modified with different dietary supplements. A total of 96 bovine enamel samples were prepared and allocated to eight groups (1-8; n = 12). Samples were eroded (120 s) in 200 ml of the following eight solutions: 1: water (control), 2: orange juice, 3: water + calcium effervescent tablet, 4: orange juice + calcium effervescent tablet, 5: water + 0.75 g acid/base regulating powder (Probase), 6: water + 0.375 g Probase, 7: orange juice + 0.75 g Probase and 8: orange juice + 0.375 g Probase. After erosion, the samples were brushed with 40 brushing strokes (load 2.5 N). Enamel wear was measured using surface profilometry after 20 and 40 cycles of erosion/abrasion respectively. Highest mean enamel wear (± SD) after 20 and 40 cycles of erosion/abrasion was observed for the unmodified orange juice (group 2) (0.605 ± 0.240 μm; 1.375 ± 0.496 μm respectively). The enamel wear in all other groups (3-8) was significantly lower (P < 0.0001 respectively) with no significant difference within these groups and compared with water (control). Erosive/abrasive enamel wear induces by orange juice and tooth brushing could be reduced significantly by modification with free available dietary supplements.
Sigma-Aldrich
Potassium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium bicarbonate, ≥99.95% trace metals basis, 99.7-100.5% dry basis
Sigma-Aldrich
Potassium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, E501, 99.5-101.0% (acidimetric)
Sigma-Aldrich
Potassium bicarbonate, ACS reagent, 99.7%, powder, crystals or granules