Merck
  • Development and in vitro evaluation of surfactant systems for controlled release of zidovudine.
Development and in vitro evaluation of surfactant systems for controlled release of zidovudine.
Journal of pharmaceutical sciences (2009-12-08)
Flávia C Carvalho, Victor H V Sarmento, Leila A Chiavacci, Mariana S Barbi, Maria P D Gremião
ABSTRACT
The development of a controlled-release dosage form of zidovudine (AZT) is of crucial importance, in view of the pharmacokinetics of its toxic activity. A suitable drug delivery system could increase AZT bioavailability, reducing its dose-dependent side effects. In this study, systems composed of polyoxypropylene (5) polyoxyethylene (20) cetyl alcohol as surfactant (S), oleic acid as oil phase (O), and water (W) were developed, as possible AZT control release systems. They were characterized by polarized light microscopy (PLM), SAXS, and rheological analysis, followed by in vitro release assay. PLM and SAXS results indicated that the mixtures of S/O/W in the proportions 55/35/10 and 55/25/20 formed microemulsion (ME) systems, while 55/20/25 formed lamellar phase. The incorporation of AZT in these systems was greater than in water or oil; moreover, AZT incorporation did not significantly change the phase behavior of the mixtures. MEs behave as Newtonian fluids in flow rheological analysis and the lamellar phase as a pseudoplastic fluid. The release profile indicated that AZT could be released in a controlled manner, since an exponential pattern governs AZT diffusion, as demonstrated by the Weibull mathematical model. These systems are potential carriers for AZT and could have advantages over conventional pharmaceutical forms.
Sigma-Aldrich
1-Hexadecanol, ≥99%
Sigma-Aldrich
1-Hexadecanol, 95%
Sigma-Aldrich
1-Hexadecanol, ReagentPlus®, 99%
Supelco
Cetyl alcohol, analytical standard
Sigma-Aldrich
Cetyl alcohol, SAJ special grade, ≥98.0%
Supelco
Cetyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Cetyl alcohol, European Pharmacopoeia (EP) Reference Standard