Merck
Biofouling control using microparticles carrying a biocide.
Biofouling (2009-11-26)
C Ferreira, R Rosmaninho, M Simoes, M C Pereira, M M S M Bastos, O C Nunes, M Coelho, L F Melo
ABSTRACT
This study presents a new technological approach to minimize the use of antimicrobial (AMB) agents and their deleterious effects, based on the principle of drug-delivery systems whereby the AMB chemicals are transported on microparticles. The efficacy of microparticles carrying the quaternary ammonium compound (QAC), benzyldimethyldodecyl ammonium chloride (BDMDAC), was assessed against Pseudomonas fluorescens in both the planktonic and the biofilm state. The microparticles were prepared using a layer-by-layer (LBL) self-assembly technique. Oppositely charged molecules of polyethyleneimine (PEI), sodium polystyrene sulfonate (PSS), and BDMDAC were assembled on polystyrene (PS) cores. BDMDAC-coated particles were observed by CryoSEM and their composition analyzed by X-ray microanalysis. Zeta potential measurements indicated that changes in surface charge were compatible with a BDMDAC/particle interaction. This biocidal carrier structure had significant stability, verified by the release of only 15% of the BDMDAC when immersed in water for 18 months. Biocidal carrier activity was evaluated by determining the survival ratio of P. fluorescens planktonic and biofilm cells after different exposure periods to BDMDAC-coated particles. Tests with biofilm cells were also performed with the free QAC. An efficient AMB effect (minimum bactericidal concentration) against suspended cells was found for a concentration of 9.2 mg l(-1) of BDMDAC on coated particles after incubation for 30 min and 6.5 mg l(-1) of BDMDAC on coated particles after 60 min. Exposure of biofilms to PS-PEI/PSS/BDMDAC (0.87 mg l(-1)) resulted in a decrease in viability of 60.5% and 66.5% of the total biofilm population for 30 and 60 min exposure times, respectively. Exposure for 60 min to 6.33 mg l(-1) and 11.75 mg l(-1) of BDMDAC in PS-PEI/PSS/BDMDAC particles promoted inactivation of 80.6% and 87.2% of the total population, respectively. The AMB effects obtained with the application of free BDMDAC were statistically similar to those promoted by the application of BDMDAC coated particles. The overall results indicate that this novel AMB strategy has potential for the control of microbial growth of planktonic cells and biofouling. Moreover, the technique allows the reuse of AMB molecules and consequently reduces the environmental risks associated with excessive use of AMB agents, thereby providing real benefits to public health.
Sigma-Aldrich
Benzyldodecyldimethylammonium bromide, ≥99.0% (AT)
Sigma-Aldrich
Benzyldimethyldodecylammonium chloride, ≥99.0% (AT)
Sigma-Aldrich
Benzyldodecyldimethylammonium bromide