Merck
Total synthesis of (+)-schweinfurthins B and E.
The Journal of organic chemistry (2009-08-25)
Joseph J Topczewski, Jeffrey D Neighbors, David F Wiemer
ABSTRACT
The first total synthesis of (+)-schweinfurthin B, a potent and differentially active cytotoxic agent, has been accomplished. Completion of the synthesis required just 16 steps in the longest linear sequence from commercially available vanillin. Key synthetic transformations included a Shi epoxidation and an efficient cascade cyclization initiated by treatment of the resulting epoxide with BF(3).OEt(2). Furthermore, use of a methyl ether as a stable protecting group for benzylic alcohols dramatically increased the efficiency of the overall sequence. The benzylic ether can be removed from this electron-rich aromatic system through oxidation with DDQ that provided the desired aldehyde intermediate in quantitative yield and shortened the synthetic sequence. Introduction of the A-ring diol in the required cis stereochemistry then became viable through a short sequence highlighted by an aldol condensation with benzaldehyde to introduce an olefin as a latent carbonyl group at the C-3 position. This synthesis established for the first time the absolute stereochemistry of the natural product, and provides access to material on a scale that will advance biological studies. The total synthesis of the closely related compound (+)-schweinfurthin E also is reported.
Sigma-Aldrich
Boron trifluoride, electronic grade, ≥99.99%
Supelco
Boron trifluoride - 1-butanol solution, ~10% in 1-butanol (∼1.3 M), for GC derivatization, LiChropur