Merck
Toxicology laboratory analysis and human exposure to p-chloroaniline.
Clinical toxicology (Philadelphia, Pa.) (2008-07-09)
Anthony F Pizon, Anna R Schwartz, Leo M Shum, Jon C Rittenberger, Darla R Lower, Spiros Giannoutsos, Mohamed A Virji, Matthew D Krasowski
ABSTRACT
p-Chloroaniline is more potent at producing methemoglobin than aniline in animal models. This case highlights the clinical presentation of an inhalation exposure to p-chloroaniline and associated laboratory analysis. An in-vitro study evaluating the metabolism of p-chloroaniline in human hepatocytes was undertaken to evaluate the metabolic fate more closely. A 20 year-old man was working at a chemical waste plant when he developed dizziness, abdominal pain, and nausea. The exam was remarkable for coma, tachycardia, cyanosis, and pulse oximetry of 75%. Arterial blood gases showed a pH 7.38, pCO(2) 41 mmHg, pO(2) 497 mmHg, bicarbonate 24 mEq/L and methemoglobin 69%. Methylene blue administration led to complete recovery without sequelae. p-Chloroaniline was later identified as the chemical involved. He denied direct contact with the chemical, but was not wearing a dust mask or respirator. GC/MS confirmed p-chloroaniline and metabolites in the patient's urine. Human hepatocytes were incubated with 100 microM p-chloroaniline for 24 hours, in both rifampicin- and vehicle only-treated cells. The cell culture medium was collected for GC/MS analysis for p-chloroaniline metabolites. Similar to the patient sample, both p-chloroaniline and p-chloroacetanilide were identified by GC/MS in hepatocytes incubated with p-chloroaniline. Neither p-chloroaniline incubated in empty cell culture nor direct GC/MS injection of p-chloroaniline generated any p-chloroacetanilide via non-enzymatic degradation. The seemingly innocuous dermal and inhalation exposure to p-chloroaniline dust can lead to life-threatening methemoglobinemia. The diagnosis can be confirmed with GC/MS analysis of the patient's urine, searching for p-chloroaniline and its primary metabolite p-chloroacetanilide.
Sigma-Aldrich
4-Chloroaniline, 98%
Supelco
4-Chloroaniline, PESTANAL®, analytical standard
Sigma-Aldrich
4-Chloroaniline, purified by sublimation, ≥99%
Supelco
p-Chloroaniline, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetaminophen Related Compound J, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4′-Chloroacetanilide, 97%