Merck
  • Validation of non-fluorescent methods to reliably detect acrosomal and plasma membrane integrity of common marmoset (Callithrix jacchus) sperm.
Validation of non-fluorescent methods to reliably detect acrosomal and plasma membrane integrity of common marmoset (Callithrix jacchus) sperm.
Theriogenology (2008-05-16)
R R Valle, C M R Valle, M Nichi, J A P C Muniz, P L Nayudu, M A B V Guimarães
ABSTRACT
Simple, rapid and stable sperm evaluation methods which have been optimized for common marmoset (Callithrix jacchus) are critical for studies involving collection and evaluation of sperm in the field. This is particularly important for new species groups such as Callitrichidae where the sperm have been little studied. Of this family, C. jacchus is the best known, and has been chosen as a model species for other members of the genus Callithrix. The fundamental evaluation parameters for sperm of any species are viability and acrosomal status. Semen samples were collected by penile vibratory stimulation. To evaluate sperm plasma membrane integrity, Eosin-Nigrosin was tested here for the common marmoset sperm to be used under field conditions. Further, a non-fluorescent stain for acrosome, the "Simple" stain, developed for domestic and wild cats, was tested on common marmoset sperm. This was compared with a fluorescent staining, Fluorescein isothiocyanate-Pisum sativum agglutinin (FITC-PSA), routinely used and validated for common marmoset at the German Primate Centre to evaluate acrosomal integrity. Results obtained with the "Simple" stain showed a marked differentiation between sperm with intact and non-intact acrosome both with and without ionophore treatment and closely correlated with results obtained with FITC-PSA. Temperature had no effect on the results with the "Simple" stain and the complete processing is simple enough to be carried out under field conditions. These findings indicated that the "Simple" stain and Eosin-Nigrosin provide rapid and accurate results for C. jacchus sperm and that those methods can be reliably used as field tools for sperm evaluation for this species.
Sigma-Aldrich
Nigrosin Stain solution, for microscopy
Sigma-Aldrich
Nigrosin water soluble, For use as a biological stain
Sigma-Aldrich
Nigrosin, certified by the Biological Stain Commission, certified by the BSC