Merck
  • Enzyme-catalysed synthesis and reactions of benzene oxide/oxepine derivatives of methyl benzoates.
Enzyme-catalysed synthesis and reactions of benzene oxide/oxepine derivatives of methyl benzoates.
Organic & biomolecular chemistry (2008-03-26)
Derek R Boyd, Narain D Sharma, John S Harrison, John F Malone, W Colin McRoberts, John T G Hamilton, David B Harper
ABSTRACT
A series of twelve benzoate esters was metabolised, by species of the Phellinus genus of wood-rotting fungi, to yield the corresponding benzyl alcohol derivatives and eight salicylates. The isolation of a stable oxepine metabolite, from methyl benzoate, allied to evidence of the migration and retention of a carbomethoxy group (the NIH Shift), during enzyme-catalysed ortho-hydroxylation of alkyl benzoates to form salicylates, is consistent with a mechanism involving an initial arene epoxidation step. This mechanism was confirmed by the isolation of a remarkably stable, optically active, substituted benzene oxide metabolite of methyl 2-(trifluoromethyl)benzoate, which slowly converted into the racemic form. The arene oxide was found to undergo a cycloaddition reaction with 4-phenyl-1,2,4-triazoline-3,5-dione to yield a crystalline cycloadduct whose structure and racemic nature was established by X-ray crystallography. The metabolite was also found to undergo some novel benzene oxide reactions, including epoxidation to give an anti-diepoxide, base-catalysed hydrolysis to form a trans-dihydrodiol and acid-catalysed aromatisation to yield a salicylate derivative via the NIH Shift of a carbomethoxy group.
Sigma-Aldrich
4-Phenyl-1,2,4-triazoline-3,5-dione, 97%
Sigma-Aldrich
Methyl benzoate, ≥98%, FCC, FG
Sigma-Aldrich
Methyl benzoate, 99%
Supelco
Methyl benzoate, analytical standard
Sigma-Aldrich
Methyl benzoate, natural, ≥98%, FCC, FG