Merck
Studying translational control in Drosophila cell-free systems.
Methods in enzymology (2007-10-05)
Fátima Gebauer, Matthias W Hentze
ABSTRACT
Classically, Drosophila cell-free translation systems have been used to study the response of the translational machinery to heat shock treatment. We and others have developed optimized Drosophila embryo and ovary extracts, and their use has expanded to the study of a variety of translational control events. These extracts recapitulate many of the aspects of mRNA translation observed in vivo and retain critical regulatory features of several translational control processes. Indeed, their use is rapidly improving our knowledge of molecular mechanisms of translational control. In this chapter we provide general guidelines and detailed protocols to obtain and use translation extracts derived from Drosophila embryos and ovaries.
Sigma-Aldrich
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
Sigma-Aldrich
Potassium acetate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Potassium acetate, meets USP testing specifications
Sigma-Aldrich
Potassium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium acetate, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium acetate, 99.98% trace metals basis
Sigma-Aldrich
Potassium acetate, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Potassium acetate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium acetate, puriss., meets analytical specification of Ph. Eur., BP, E261, 99-101%
Sigma-Aldrich
Potassium acetate, ACS reagent, ≥99.0%