Merck
  • Hyphenation of infrared spectroscopy to liquid chromatography for qualitative and quantitative polymer analysis: degradation of poly(bisphenol A)carbonate.
Hyphenation of infrared spectroscopy to liquid chromatography for qualitative and quantitative polymer analysis: degradation of poly(bisphenol A)carbonate.
Journal of chromatography. A (2006-06-01)
Leon Coulier, Erwin Kaal, Thomas Hankemeier
ABSTRACT
Hyphenation of infrared spectroscopy (IR) to liquid chromatography (LC) has been applied to study chemical changes in poly(bisphenol A)carbonate (PC) as a result of degradation. Especially coupling of LC to FTIR through solvent elimination is a sensitive approach to identify changes in functionality observed in the LC chromatograms as has been demonstrated by coupling of liquid chromatography under critical conditions (LCCC) to IR. Furthermore, an example is shown in which two-dimensional liquid chromatography, i.e. LCCC x SEC, was coupled to IR by means of a flow cell. This resulted in data sets containing most probably valuable data, but extracting relevant information from these large data sets is not straightforward at all. Therefore, multivariate data analysis (MVDA) of SEC-FTIR data was used to extract relevant data from large data sets. This approach revealed chemical differences due to degradation that could not be detected by other means. Spectral features could be identified that allowed to quantitatively predict the degradation of poly(bisphenol A)carbonate as a function of degradation conditions.
Product Description
Supelco
Poly(Bisphenol A carbonate), analytical standard
Supelco
Poly(Bisphenol A carbonate)
Sigma-Aldrich
Poly(Bisphenol A carbonate), melt index 10-12 g/10 min (300°C/1.2kg)
Sigma-Aldrich
Poly(Bisphenol A carbonate), average Mw ~45,000 by GPC