Merck
[Analysis of phenmedipham in agricultural products by HPLC].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2006-01-31)
Tsuyoshi Imazawa, Tomonari Iida, Nobuhiro Matsuno, Fumiaki Kato, Takeshi Ito, Kumiko Sasaki
ABSTRACT
An analytical method was developed for the determination of phenmedipham (PM) in agricultural products using reversed-phase high-performance liquid chromatography with UV detection. A sample was extracted with acetonitrile, and the acetonitrile layer was separated by salting-out. The acetonitrile phase was isolated and evaporated. The extract was dissolved in diethyl ether-hexane (1 : 1), and then cleaned up on a Florisil column. The column was washed with diethyl ether-hexane (1 : 1), and PM was eluted with acetone-hexane (3 : 7), and the eluate was evaporated. The residue was dissolved in acetone-hexane (2 : 8), and the sample solution was cleaned up on SAX/PSA cartridge. The SAX/PSA cartridge was washed with acetone-hexane (2 : 8), and PM was eluted with acetone-hexane (3 : 7). If required, the eluate of the Florisil column was cleaned up with SAX/PSA and ENVI-Carb/ NH2 cartridges. The SAX/PSA cartridge was washed with acetone-hexane (2 : 8), and connected to be ENVI-Carb/NH2 cartridge. The cartridges were washed with acetone-hexane (3 : 7), and then the SAX/PSA cartridge was removed. PM was eluted with acetonitrile-toluene (3 : 1) from the ENVI-Carb/NH2 cartridge. PM in the eluate was separated isocratically on an ODS column (4.6 mm i.d. x 150 mm, 5 microm) using acetonitrile-water (6 : 4) as a mobile phase (flow-rate 1.0 mL/min, temp. 40 degrees C), with monitoring at 235 nm. The calibration curve was linear from 0.005 microg/mL to 10 microg/mL of PM. The recoveries of PM from eight kinds of agricultural products spiked at levels of 0.1 and 0.02 microg/g were 80.8-98.7%. The determination limit was 0.01 microg/g.
Product Description
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Phenmedipham, PESTANAL®, analytical standard