Merck
Origin of 2-ethylhexanol as a VOC.
Environmental pollution (Barking, Essex : 1987) (2005-08-30)
Sandro Nalli, Owen J Horn, Adam R Grochowalski, David G Cooper, Jim A Nicell
ABSTRACT
2-Ethylhexanol has been identified as a volatile organic compound (VOC) that contributes to the deterioration of indoor air quality. Plasticizers are common components of dust and building materials and are shown to be degraded by a variety of bacteria and fungi to produce 2-ethylhexanol and other metabolites. Of these, the 2-ethylhexanol has significant volatility and was observed in appreciable quantities. The degree to which 2-ethylhexanol is observed as a VOC in air samples would be limited by the fact that many of the microorganisms that are capable of producing this compound are also able to oxidize it to 2-ethylhexanoic acid, which is much less volatile. It is argued that an abiotic degradation mechanism of plasticizers that results in the generation of 2-ethylhexanol is unlikely and, if this did occur, other metabolites should have been observed. Thus, the microbial degradation of plasticizers is the most likely source of 2-ethylhexanol in indoor air.
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99.6%
Sigma-Aldrich
Titanium(IV) 2-ethylhexyloxide, 95%
Sigma-Aldrich
2-Ethyl-1-hexanol, puriss., ≥99.0% (GC)
Supelco
2-Ethyl-1-hexanol, analytical standard
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99%, FG