Merck
  • Urinary excretion profiles of two major triazolam metabolites, alpha-hydroxytriazolam and 4-hydroxytriazolam.
Urinary excretion profiles of two major triazolam metabolites, alpha-hydroxytriazolam and 4-hydroxytriazolam.
Journal of analytical toxicology (2005-06-25)
Kenji Tsujikawa, Kenji Kuwayama, Hajime Miyaguchi, Tatsuyuki Kanamori, Yuko Iwata, Hiroyuki Inoue, Tohru Kishi
ABSTRACT
The objective of this study was to examine urinary excretion profiles of two major triazolam metabolites, alpha-hydroxytriazolam (alpha-OHTRZ) and 4-hydroxytriazolam (4-OHTRZ) in humans. Urine samples were collected from three healthy male volunteers who had been previously administered single 0.25- and 0.5-mg doses of triazolam 24 h and 48 h, respectively, before sample collection. After enzymatic hydrolysis and extraction, each sample was analyzed by liquid chromatography-mass spectrometry. alpha-OHTRZ was rapidly excreted, with the maximum concentrations appearing in the first or second sample collected after ingestion, with the majority of the drug being excreted within 12 h. Meanwhile, 4-OHTRZ was excreted more slowly than alpha-OHTRZ. The alpha-OHTRZ/4-OHTRZ ratios were initially greater than 19.7, then decreased rapidly, reaching a nearly constant value for times in excess of 12 h.
Sigma-Aldrich
α-Hydroxytriazolam
Supelco
α-Hydroxytriazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®