Merck
  • Triisobutylaluminium and diisobutylaluminium hydride as molecular scalpels: the regioselective stripping of perbenzylated sugars and cyclodextrins.
Triisobutylaluminium and diisobutylaluminium hydride as molecular scalpels: the regioselective stripping of perbenzylated sugars and cyclodextrins.
Chemistry (Weinheim an der Bergstrasse, Germany) (2004-06-24)
Thomas Lecourt, Alexandre Herault, Alan J Pearce, Matthieu Sollogoub, Pierre Sinaÿ
ABSTRACT
To explain the remarkable regioselective de-O-benzylating properties of diisobutylaluminium hydride (DIBAL-H) and triisobutylaluminium (TIBAL) towards polybenzylated sugars or cyclodextrins, we propose a plausible mechanistic rationale critically involving the kinetic formation of a product-generating 2:1 Al-benzylated sugar complex. For the reaction to occur, one pair of adjacent oxygen atoms should first be able to form a chelation complex with the first equivalent of aluminium reagent, either a highly fluxional complex with tetracoordinate aluminium species or a pentacoordinate one. The second equivalent then induces the regioselectivity of the de-O-alkylation by coordinating preferentially to one of the oxygen atoms of the selected pair.
Sigma-Aldrich
Diisobutylaluminum hydride, reagent grade
Sigma-Aldrich
Diisobutylaluminum hydride solution, 25 wt. % in toluene
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in hexanes
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in heptane
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in toluene
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in cyclohexane
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in methylene chloride