Merck
  • Suggested improvements to the standard filter paper assay used to measure cellulase activity.
Suggested improvements to the standard filter paper assay used to measure cellulase activity.
Biotechnology and bioengineering (2003-04-04)
Guillermo Coward-Kelly, Cateryna Aiello-Mazzari, Sehoon Kim, Cesar Granda, Mark Holtzapple
ABSTRACT
Two suggestions can be found in the literature to improve the reproducibility of the Mandels' filter paper assay: add supplemental cellobiase and increase the boiling time for color development. Here we provide data that strongly supports adding supplemental cellobiase. Adding supplemental cellobiase increased assay response by 56%. Cellulases from different sources have different cellobiase activities, which would cause significant variation in the assay response. There is no need for additional boiling time-5 minutes is sufficient. For maximum reproducibility, it is essential that the water bath vigorously boil so that temperature excursions are minimized.
Sigma-Aldrich
3,5-Dinitrosalicylic acid, used in colorimetric determination of reducing sugars
Sigma-Aldrich
3,5-Dinitrosalicylic acid, 98%