Merck
Im-trityl protection of histidine.
Journal of peptide science : an official publication of the European Peptide Society (1999-10-03)
S J Harding, J H Jones, A N Sabirov, V V Samukov
ABSTRACT
A rational attempt to prepare FmocHis(piTrt)OH regiospecifically gave in fact the well-known tau-trityl isomer, and experiments with model systems indicate that the prospects for access to pi-trityl histidine derivatives, which would be of great value for the racemization-free synthesis of histidine-containing peptides, are poor.
Sigma-Aldrich
Trityl chloride, 97%
Sigma-Aldrich
Trityl chloride, purum, ≥97.0% (HPLC), ≥97.0% (AT)