Merck
  • New Dihydropyridine Derivative Attenuates NF-κB Activation via Suppression of Calcium Influx in a Mouse BV-2 Microglial Cell Line.
New Dihydropyridine Derivative Attenuates NF-κB Activation via Suppression of Calcium Influx in a Mouse BV-2 Microglial Cell Line.
The Tohoku journal of experimental medicine (2024-04-04)
Kota Sato, Yuto Sasaki, Michiko Ohno-Oishi, Kuniyuki Kano, Junken Aoki, Kosuke Ohsawa, Takayuki Doi, Hiroyuki Yamakoshi, Yoshiharu Iwabuchi, Chihiro Kawano, Yoshiyuki Hirata, Toru Nakazawa
ABSTRACT
Activated microglia contribute to many neuroinflammatory diseases in the central nervous system. In this study, we attempted to identify an anti-inflammatory compound that could suppress microglial activation. We performed high-throughput screening with a chemical library developed at our institute. We performed a luciferase assay of nuclear factor-kappa B (NF-κB) reporter stable HT22 cells and identified a compound that was confirmed to inhibit the anti-inflammatory response in BV2 microglial cells. The selected dihydropyridine derivative can suppress the expression response of interleukin-1β (IL-1β), interleukin-6 (IL-6), and tumor necrosis factor (TNF), as well as NF-κB phosphorylation and nuclear translocation, and reduce the intracellular calcium level. Thus, our identified compound has a potential role in suppressing microglial activation and may contribute to the development of a new therapeutic molecule against neuroinflammatory diseases.
Sigma-Aldrich
Anti-goat IgG (H+L)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody